4-Deoxypyridoxine hydrochloride ≥98%, crystalline solid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:148-51-6
:
Formula:C₈H₁₁NO₂
MDL Number:MFCD00051296
:189.64 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98%
- λmax:286 nm