Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
X-Gal
X-Gal staining kit
X-Gal staining kit
X-Gal staining kit
Catalog # OZBIGX10003
Supplier:
OZ BIOSCIENCES
CAS Number:
Expand image
X-Gal staining kit
Catalog # OZBIGX10003
Supplier:
OZ BIOSCIENCES
CAS Number:
Specifications
Fp:
1 KIT
Seq:
0075
Specifications
About this item
Product Details
Documentation
:
:
408.63 g/mol
CAS Number:
7240-90-6
Formula:
C₁₄H₁₅BrClNO₆
MDL Number:
MFCD00005666
Product Details
Documentation