Order Entry
Norway
ContactUsLinkComponent
Sodium azide 0.05% in water, Supelco®
Sodium azide 0.05% in water, Supelco®
Catalog # RTC000068-1L
Supplier:  Merck
CAS Number:  
Sodium azide 0.05% in water, Supelco®
Catalog # RTC000068-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :65.01 g/mol
CAS Number:26628-22-8
Formula:NaN₃
MDL Number:MFCD00003536