D-(+)-Raffinose pentahydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17629-30-0
:
Formula:C₁₈H₃₂O₁₆·5H₂O
MDL Number:MFCD00071590
:594.52 g/mol
Merck Index:14,08089
Boiling Pt:78…82 °C
Storage Temperature:Ambient
EINECS:208-146-9
Specification Test Results
- Identification (FTIR):Conforms
- Residue on Ignition:≤0.05%
- Loss on Drying:≤15.3%
- Optical Rotation (c=4 in H₂O and NH₃):+104.9 ±1.3°
- Arsenic (As):≤0.5 ppm
- Heavy Metal (Cu):≤10 ppm
- Iron (Fe):≤5 ppm