Glycine 99+%, Ultrapure for molecular biology
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Merck Index:14,04491
Boiling Pt:181 °C (1013 hPa)
Melting Pt:∼ 245 °C (dec.)
Density:1.595
Storage Temperature:Ambient
EINECS:200-272-2
Specification Test Results
- Assay (Unspecified):≥99.0%
- Assay (Unspecified) - Hydrolysable Substances:Passes Test
- Assay (Unspecified) - Readily Carbonisable Substances:Passes Test
- Chloride Content:≤50 ppm
- Identification (FTIR):Conforms
- Residue on Ignition:≤0.1%
- Impurity Content Sulphate (SO₄):≤50 ppm
- lambda-max (UV-Vis) - (1 M, 1 cm ≤0.05):260 nm
- Arsenic (As):≤1 ppm
- Barium (Ba):≤1 ppm
- Cadmium (Cd):≤1 ppm
- Copper (Cu):≤1 ppm
- Iron (Fe):≤1 ppm
- Lead (Pb):≤1 ppm
- Magnesium (Mg):≤1 ppm
- Manganese (Mn):≤1 ppm
- Zinc (Zn):≤1 ppm