Order Entry
Norway
ContactUsLinkComponent
Anti-SAP6 Mouse Monoclonal Antibody [clone: X63] Saporin
Anti-SAP6 Mouse Monoclonal Antibody [clone: X63] Saporin
Supplier:  Biosensis
Certificates
Anti-SAP6 Mouse Monoclonal Antibody [clone: X63] Saporin
Supplier:  Biosensis

Specifications

  • Catalog No:
  • BSENM-1792-25-IT
  • BSENM-1792-50-IT
  • BSENM-1792-100-IT
  • Antigen symbol:
  • SAP6
  • SAP6
  • SAP6
  • Antigen name:
  • Non-specific Control IgG;Mouse Monoclonal Antibody (Saporin)
  • Non-specific Control IgG;Mouse Monoclonal Antibody (Saporin)
  • Non-specific Control IgG;Mouse Monoclonal Antibody (Saporin)
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Saporin
  • Saporin
  • Saporin
  • Clone:
  • X63
  • X63
  • X63
  • Reactivity:
  • Species independent
  • Species independent
  • Species independent
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID #P20656
  • UniprotID #P20656
  • UniprotID #P20656
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • Amino acid number:
  • 299 amino acids
  • 299 amino acids
  • 299 amino acids
  • Form:
  • Lyophillised
  • Lyophillised
  • Lyophillised
  • Molecular weight:
  • 30 kDa
  • 30 kDa
  • 30 kDa
  • Purification:
  • Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
  • Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
  • Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • Reconstitute in sterile-filtered, ultrapure water
  • Reconstitute in sterile-filtered, ultrapure water
  • Reconstitute in sterile-filtered, ultrapure water
  • Storage temperature:
  • 2…8 °C (lyophilised);<–70 °C (liquid)
  • 2…8 °C (lyophilised);<–70 °C (liquid)
  • 2…8 °C (lyophilised);<–70 °C (liquid)
  • Shipping temperature:
  • 25 °C
  • 25 °C
  • 25 °C

Specifications