Order Entry
Norway
ContactUsLinkComponent
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Catalog # VWRS25642.550
Supplier:  VWR Collection
CAS Number:  
undefined
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Catalog # VWRS25642.550
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • IR Spectrum:
    Passes test
  • Acidity (at filling):
    Max. 0.0005 meq/g
  • Boiling point:
    79 - 80 °C
  • Colouration:
    Max. 10 APHA
  • Density (20/4):
    0.804 - 0.807
  • Density (20/20):
    0.805 - 0.808
  • Acetone:
    Max. 0.05 %
  • Butan-2-ol:
    Max. 0.07 %
  • Methanol:
    Max. 0.05 %
  • 2-Methylpropan-2-ol:
    Max. 0.1 %
  • Non-volatile residue:
    Max. 10 ppm
  • Substances reducing KMnO4 (as O):
    Max. 3 ppm
  • Water:
    Max. 0.1 %
  • Al (Aluminium):
    Max. 0.2 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.05 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.05 ppm
  • Mn (Manganese):
    Max. 0.02 ppm
  • Na (Sodium):
    Max. 0.5 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Sr (Strontium):
    Max. 0.02 ppm
  • Zn (Zinc):
    Max. 0.1 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

About this item

 :
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C (DIN 51755 Part 1)
Storage Temperature:Ambient
EINECS:201-159-0
UN:1193
ADR:3,II
 :72.11 g/mol
REACH:01-2119457290-43
 :GHS02
 :GHS07
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648