2,4-Diamino-6-hydroxypyrimidine
Catalog # CAYM81260-1
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:126.12 g/mol
Merck Index:13,03001
Boiling Pt:373 °C (1013 hPa)
Melting Pt:285…286 °C
EINECS:200-254-4
CAS Number:56-06-4
:MFCD08528919
Formula:C₄H₆N₄O
MDL Number:MFCD00006098