Perylene ≥99.0%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:198-55-0
:
Formula:C₂₀H₁₂
MDL Number:MFCD00004142
:252.32 g/mol
Merck Index:12,07325
Boiling Pt:443 °C (1013 hPa)
Melting Pt:276…279 °C (lit.)
Density:1.362 g/cm³ (20 °C)
Storage Temperature:Room Temperature
EINECS:205-900-9
Specification Test Results
- Appearance:Gold to orange crystalline powder
- Infrared spectrum:Conforms to structure.
- Elemental analysis:Carbon 94.2% - 96.2%
- High pressure liquid:99.0% (minimum)