Nickel (II) oxide, black 76% Ni
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1313-99-1
:
Formula:NiO
MDL Number:MFCD00011145
:74.69 g/mol
Merck Index:13,06540
Melting Pt:1960 °C
Density:6,72 g/cm³ (20 °C)
EINECS:215-215-7
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark grey-black Powder
- Assay:75 to 77 % (Ni)