Order Entry
Norway
ContactUsLinkComponent
Cobalt(II) chloride, anhydrous for synthesis, Sigma-Aldrich®
Corrosive
Health
Irritant
Environment
Cobalt(II) chloride, anhydrous for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Health
Irritant
Environment
Cobalt(II) chloride, anhydrous for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    735 °C
  • Molar Mass:
    129.83 g/mol
  • Boiling Point:
    1049 °C (1013 hPa)
  • Density:
    3.36 g/cm3 (25 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number