


Cobalt(II) chloride, anhydrous 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7646-79-9
:
Formula:Cl₂Co
MDL Number:MFCD00010938
:129.84 g/mol
Merck Index:12,02498
Boiling Pt:1049 °C (1013 hPa)
Melting Pt:735 °C
Density:1,6795 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:231-589-4
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Blue Powder
- Titration Complexometric:≥96.0 %
- Iron (Fe):≤500 ppm
- Lead (Pb):≤100 ppm
- Nickel (Ni):≤2000 ppm