Order Entry
Norway
ContactUsLinkComponent
Zinc chloride 98-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Zinc chloride 98-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Katalog # 14422-25KG
CAS Number:  
Zinc chloride 98-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Katalog # 14422-25KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 kg
  • Seq:
    4286
  • Pk Info:
    1×25 kg

Specifications

Specification Test Results

  • Assay:
    98.0 - 100.5 %
  • Oxide chloride (as ZnO):
    complying
  • pH (10 %, 20°C):
    4.6 - 5.5
  • Ammonium (NH₄):
    Max. 300 ppm
  • Arsenic (As):
    Max. 0.0002 %
  • Calcium (Ca):
    Max. 0.01 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.02 %
  • Magnesium (Mg):
    Max. 0.01 %
  • Sodium (Na):
    Max. 0.01 %
  • Lead (Pb):
    Max. 0.005 %
  • Total N:
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 200 ppm
  • Vapor Pressure:
    1 mmHg ( 428 °C)
  • Al, Ca, Fe, Mg and heavy metals:
    complying
  • Alkali and alkaline earth:
    Max. 1.0 %
  • Residual solvents:
    complying

Specification Test Results

About this item

 :
 :136.3 g/mol
Merck Index:13,10185
Boiling Pt:732 °C (1013 hPa)
Melting Pt:287…304 °C (1.013 hPa)
Density:2.93 g/cm³ (22 °C)
Storage Temperature:Ambient
EINECS:231-592-0
UN:2331
ADR:8,III
 :GHS07
 :GHS05
 :GHS09
CAS Number:7646-85-7
Formula:ZnCl₂
MDL Number:MFCD00011295