Order Entry
Norway
ContactUsLinkComponent
Silver chromate ≥99%
Silver chromate ≥99%
Catalog # 40107.14
CAS Number:  
Silver chromate ≥99%
Catalog # 40107.14
CAS Number:  

Specifications

  • Pk:
    25 g

Specifications

About this item

 :
 :331.73 g/mol
Merck Index:14,8510
Boiling Pt:1550 °C (1013 hPa)
Density:5.63
Storage Temperature:Ambient
EINECS:232-043-8
UN:1479
ADR:5.1,III
 :GHS08
 :GHS03
 :GHS07
CAS Number:7784-01-2
Formula:Ag₂CrO₄
MDL Number:MFCD00003402