ESD safety shoes, lace-up, Basic, jo_Colour Orange Low, Jori
Supplier: ELTEN
Certificates
About this item
Safety shoes made from hydrophobised, quick drying, tear resistant textile upper combined with reflective materials, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU (New Basic) sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
- Climate-regulating, breathable textile lining
- Full length inlay sole
- PU scuff cap protects the upper material
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/orange
Certifications: EN 61340 (ESD); EN ISO 20345; S3, SRC.
Specifications
- Catalog No:
- ELTN12941/38
- ELTN12941/37
- ELTN12941/48
- ELTN12941/47
- ELTN12941/36
- ELTN12941/42
- ELTN12941/41
- ELTN12941/40
- ELTN12941/39
- ELTN12941/46
- ELTN12941/45
- ELTN12941/44
- ELTN12941/43
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001