Order Entry
Norway
ContactUsLinkComponent
Dysprosium(III) oxide, Sigma-Aldrich®
Dysprosium(III) oxide, Sigma-Aldrich®
Catalog # 1.12151.0010
Supplier:  Merck
CAS Number:  
Dysprosium(III) oxide, Sigma-Aldrich®
Catalog # 1.12151.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    2340 °C
  • Molar Mass:
    372.99 g/mol
  • Density:
    7.81 g/cm3 (20 °C)

Specification Test Results

About this item

 :
Merck Index:14,03482
Melting Pt:∼2400 °C
Density:7.81 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :373 g/mol
EINECS:215-164-0
CAS Number:1308-87-8
Formula:Dy₂O₃
MDL Number:MFCD00010986