Dysprosium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1308-87-8
:
Formula:Dy₂O₃
MDL Number:MFCD00010986
Merck Index:14,3482
Melting Pt:2340 °C
Density:7.81
:373 g/mol
Storage Temperature:Ambient
EINECS:215-164-0