Order Entry
Norway
ContactUsLinkComponent
6-Methylindole 99%
Irritant
6-Methylindole 99%
Certificates
undefined
Irritant
6-Methylindole 99%

Specification Test Results

  • Appearance:
    Clear yellow to light brown Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6060 to 1.6080 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information