5,6-Dimethoxyindole
Glass bottle for solids, 1 g
Catalog # MOLEM82103323
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:14430-23-0
- Formula:C₁₀H₁₁NO₂
- MDL Number:MFCD00005675
- Molecular Weight:177.2 g/mol
- EINECS:238-402-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



