Order Entry
Norway
ContactUsLinkComponent
5-Fluoroindole 98%
Irritant
5-Fluoroindole 98%
Certificates
undefined
Irritant
5-Fluoroindole 98%

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    44 to 48 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information