Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1'-Carbonyldiimidazole
1,1'-Carbonyldiimidazole
1,1'-Carbonyldiimidazole
1,1'-Carbonyldiimidazole
RESERES1360C-A103X
:
RESEARCH ORGANICS
:
1,1'-Carbonyldiimidazole
RESERES1360C-A103X
:
RESEARCH ORGANICS
:
Pack type:
Glass bottle for solids
Pk:
50 g
Seq:
3368
:
:
162.15 g/mol
:
14,01819
:
111…116 °C
:
Ambient
:
208-488-9
:
3263
:
8,II
:
GHS05
:
GHS07
:
530-62-1
:
C₇H₆N₄O
:
MFCD00005286