1,1'-Carbonyldiimidazole
Catalog # MOLE25890016-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:162.15 g/mol
Merck Index:14,01819
Melting Pt:111…116 °C
Storage Temperature:Ambient
EINECS:208-488-9
UN:3263
ADR:8,II
:GHS05
:GHS07
CAS Number:530-62-1
Formula:C₇H₆N₄O
MDL Number:MFCD00005286