Order Entry
Norway
ContactUsLinkComponent
 
Diallyl disulphide
Catalog # MOLE33421999-100G
Supplier:  Molekula
CAS Number:  
Diallyl disulphide
Catalog # MOLE33421999-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :146.28 g/mol
Boiling Pt:198 °C (1013 hPa)
Density:1.0237 g/cm³ (15 °C)
Storage Temperature:Refrigerator
EINECS:218-548-6
UN:2810
ADR:6.1,III
 :GHS07
CAS Number:2179-57-9
Formula:C₆H₁₀S₂
MDL Number:MFCD00008656