Amidosulphonic acid, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:5329-14-6
:
Formula:H₃NO₃S
MDL Number:MFCD00011603
:97.09 g/mol
Merck Index:13,09006
Boiling Pt:247 °C (1013 hPa)
Melting Pt:205 °C
Density:2.15 g/cm³ (20 °C)
UN:2967
ADR:8,III
:M-Clarity™ quality level = MQ300
EINECS:226-218-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:213 g/l (20 °C)
- Melting Point:205 °C (decomposition)
- Molar Mass:97.08 g/mol
- Bulk Density:600 kg/m3
- Density:2.13 g/cm3 (20 °C)
- pH:1.2 (10 g/l, H2O)
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_100103
SDS
ProductPage_100103 ProductPage_100103