Amidosulphonic acid
Katalog # HACH107399
Supplier: Hach
CAS Number:
Specifications
- Pk:100
- Seq:0100
Specifications
About this item
:
:97.09 g/mol
Merck Index:13,09006
Boiling Pt:247 °C (1013 hPa)
Melting Pt:205 °C
Density:2.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-218-8
UN:2967
ADR:8,III
:GHS07
CAS Number:5329-14-6
Formula:H₃NO₃S
MDL Number:MFCD00011603