Order Entry
Norway
ContactUsLinkComponent
Ammonium sulphate ≥99.0%, Reagent Grade
Ammonium sulphate ≥99.0%, Reagent Grade
Certificates
Ammonium sulphate ≥99.0%, Reagent Grade

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying
  • Iron (Fe):
    Max. 10 ppm
  • Lead (Pb):
    Max. 3 ppm
  • Selenium (Se):
    Max. 5 ppm

Specification Test Results

Related Information