Ammonium sulphate ≥99%, white crystalline powder, Enzyme Grade
:7783-20-2
:
:(NH₄)₂SO₄
:MFCD00003391
:132.14 g/mol
:13,00559
:336…339 °C
:1.78 g/cm³ (20 °C)
:231-984-1
:01-2119455044-46
:GHS07
:Ambient
- Purity:≥99%
- Heavy Metals:≤0,5 ppm (as Pb)
- pH:5 - 6 (5% aq soln)
- Protease, DNase, RNase:Not detected