Ammonium carbonate ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:506-87-6
:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890
:96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C (dec.)
Density:1,5 g/cm³ (20 °C)
EINECS:208-058-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder or crystals and/or chunks
- Non-volatile matter:≤0.01 %
- Titration excess HCl:≥30.0 % (NH3)
- Chloride (Cl):≤5 ppm
- Iron (Fe):≤5 ppm
- Heavy metals (ICP-OES):≤5 ppm
- Sulfur compounds (as SO4):≤0.002 %
- Insoluble matter:≤0.005 %