Ammonium chloride
Catalog # MOLE24838060-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:53.49 g/mol
Merck Index:13,00510
Melting Pt:335 °C
Density:1.5256 g/cm³ (20 °C)
EINECS:235-186-4
:GHS07
Storage Temperature:Ambient
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420