Order Entry
Norway
ContactUsLinkComponent
Tetrapropylammonium bromide, Supelco®
Tetrapropylammonium bromide, Supelco®
Catalog # 88103-10G
Supplier:  Merck
CAS Number:  
undefined
Tetrapropylammonium bromide, Supelco®
Catalog # 88103-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :266.27 g/mol
Melting Pt:265…274 °C
 :M-Clarity™ quality level = MQ100
EINECS:217-727-6
CAS Number:1941-30-6
Formula:C₁₂H₂₈BrN
MDL Number:MFCD00011840