
Tetrapentylammonium bromide, Supelco®
Catalog # 87997-10G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:378.48 g/mol
Melting Pt:103 °C
Density:1.0662 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
EINECS:212-756-0
:GHS07
CAS Number:866-97-7
Formula:C₂₀H₄₄BrN
MDL Number:MFCD00011856