Order Entry
Norway
ContactUsLinkComponent
Tetraoctylammonium bromide, Supelco®
Tetraoctylammonium bromide, Supelco®
Catalog # 87994-5G
Supplier:  Merck
CAS Number:  
undefined
Tetraoctylammonium bromide, Supelco®
Catalog # 87994-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :546.8 g/mol
Melting Pt:~ 96 °C
 :M-Clarity™ quality level = MQ100
Flash Pt:100 °C
EINECS:238-936-9
CAS Number:14866-33-2
Formula:C₃₂H₆₈BrN
MDL Number:MFCD00011863