Tetrabutylammonium bromide, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1643-19-2
:
Formula:C₁₆H₃₆BrN
MDL Number:MFCD00011633
:322.37 g/mol
Melting Pt:100…103 °C
Density:1.148 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:216-699-2