Order Entry
Norway
Orders LinkContactUsLinkComponent
Bismuth(III) oxide, Supelco®

Bismuth(III) oxide, Supelco®

Irritant
Supplier:   Merck
Bismuth(III) oxide, Supelco®
Supplier:  Merck
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Bismuth Trioxide, Dibismuth trioxide
  • CAS Number:1304-76-3
  • Formula:BiO₃
  • MDL Number:MFCD00003462
  • Molecular Weight:465.96 g/mol
  • Merck Index:13,01273
  • Boiling Pt:1890 °C (1013 hPa)
  • Melting Pt:825 °C
  • Density:8.9 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:215-134-7

Documents

Specifications for Product Family

Specification Test Results

  • Melting Point:
    820 °C
  • Molar Mass:
    465.95 g/mol
  • Bulk Density:
    1000 kg/m3
  • Boiling Point:
    1890 °C
  • Density:
    8.93 g/cm3 (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - kr 3 820,00
    kr 3 820,00
    1 * 1 kg
       kr 3 820,00

  • Fibre carton
    25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.