
Bismuth(III) oxide 99.9%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1304-76-3
:
Formula:BiO₃
MDL Number:MFCD00003462
:465.96 g/mol
Merck Index:13,01273
Boiling Pt:1890 °C (760 mmHg)
Melting Pt:825 °C
Density:8.9 g/cm³
EINECS:215-134-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow Powder
- Loss on drying:≤0.2 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤10 ppm
- Bismuth (Bi):89.5 to 89.9 %
- Sodium (Na):≤50 ppm