

(3-Aminopropyl)triethoxysilane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:919-30-2
:
Formula:H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MDL Number:MFCD00008207
:221.37 g/mol
Boiling Pt:217 °C (760 mmHg)
Melting Pt:–70 °C
Density:0.942 g/cm³
Flash Pt:96 °C
Storage Temperature:Ambient
EINECS:213-048-4
UN:2735
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4200 to 1.4220 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤25 APHA