orto-Borsyre ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Merck Index:14,1336
Boiling Pt:185 °C (1013 hPa)
Melting Pt:∼ 185 °C (dec.)
Density:1,435
:61.83 g/mol
Storage Temperature:Romtemperatur
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08