Aluminium oxide, Durmax® for metallography, 48 hours
Catalog # 20983.262
Supplier: ALUMINES DURMAX
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:101.96 g/mol
Merck Index:13,00355
Boiling Pt:2980 °C (1013 hPa)
Melting Pt:2045 °C
Density:3.94 g/cm³ (20 °C)
EINECS:215-691-6
REACH:01-2119529248-35
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424