0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminiumoksid
Aluminiumoksid substrate 10x10x0.5 mm, polished one side A plane
Aluminiumoksid substrate 10x10x0.5 mm, polished one side A plane
:
Thermo Fisher Scientific
Aluminiumoksid substrate 10x10x0.5 mm, polished one side A plane
:
Thermo Fisher Scientific
:
1344-28-1
:
11092-32-3
:
Al₂O₃
:
MFCD00003424
:
101.96 g/mol
:
13,00355
:
2980 °C (1013 hPa)
:
2045 °C
:
3,94 g/cm³ (20 °C)
:
Romtemperatur
:
215-691-6
:
01-2119529248-35
msds_ALFA038498
Thermo Scientific Acroseal 25 Years