Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium oxide
Aluminium oxide ≥99% (metals basis)
Aluminium oxide ≥99% (metals basis)
:
Thermo Fisher Scientific
Aluminium oxide ≥99% (metals basis)
:
Thermo Fisher Scientific
:
1344-28-1
:
11092-32-3
:
Al₂O₃
:
MFCD00003424
:
101.96 g/mol
:
14,356
:
2980 °C
:
2045 °C
:
3.965
:
Ambient
:
215-691-6
:
01-2119529248-35
msds_ALFA012553
Thermo Scientific Acroseal 25 Years