Aluminium oxide 99.99% (trace metals basis), extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424
:101.96 g/mol
Merck Index:13,00355
Boiling Pt:2977 °C (760 mmHg)
Melting Pt:2030 °C
Density:3.97 g/cm³
Storage Temperature:Ambient
EINECS:215-691-6
REACH:01-2119529248-35
Specification Test Results
- Appearance:White Powder
- Total trace metal impurities:≤0.01 %