Boron tribromide 99.9%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-33-4
:
Formula:BBr₃
MDL Number:MFCD00011312
:250.52 g/mol
Merck Index:13,01337
Boiling Pt:91.2 °C (760 mmHg)
Melting Pt:–46 °C
Density:2.65 g/cm³
Storage Temperature:Ambient
EINECS:233-657-9
UN:2692
ADR:8,I
:GHS06
:GHS05
Specification Test Results
- Appearance:Clear amber Liquid
- Assay:≥99.85 % (By difference)
- Titration:≥99.2 %
- Sulfur (S):≤100 ppm
- Heavy metals (as Pb):≤100 ppm
- Impurity:≤0.13 % Free bromine (Br2)