Order Entry
Norway
Orders LinkContactUsLinkComponent
Calcium hydroxide 95%

Calcium hydroxide 95%

Corrosive
Irritant
Calcium hydroxide 95%
Certificates
Calcium hydroxide 95%
Calcium hydroxide 95%

3 Options Available Below

Product Details & Documents
Synonyms:  Slaked lime, Calcium dihydroxide
  • CAS Number:1305-62-0
  • Formula:Ca(OH)₂
  • MDL Number:MFCD00010901
  • Molecular Weight:74.09 g/mol
  • Merck Index:14,1673
  • Melting Pt:550 °C
  • Density:2.24
  • Storage Temperature:Ambient
  • EINECS:215-137-3
  • UN:3262
  • ADR:8,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.