Calcium hydroxide 95+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1305-62-0
:
Formula:Ca(OH)₂
MDL Number:MFCD00010901
:74.09 g/mol
Merck Index:13,01675
Boiling Pt:2850 °C
Melting Pt:580 °C (dec.)
Density:2,24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-137-3
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White Powder
- Titration with HCl:≥95.0 % (Ca(OH)2)
- Chloride (Cl):≤300 ppm
- Iron (Fe):≤500 ppm
- Magnesium (Mg):≤5000 ppm
- Potassium (K):≤500 ppm
- Sodium (Na):≤500 ppm
- Strontium (Sr):≤500 ppm
- Heavy metals (ICP-OES):≤30 ppm
- Sulfur compounds (as SO4):≤0.1 %
- Insoluble matter:≤0.03 % (in HCl)