Potassium dihydrogen phosphate ≥99.999% (metals basis), Puratronic® (Si ≤ 10 ppm)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Merck Index:14,7659
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.338
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
:GHS07