Order Entry
Norway
Orders LinkContactUsLinkComponent
Potassium nitrate

Potassium nitrate

Plastic bottle for solids, 5 kg
Catalog # SWEDKA580412-5
Oxidizing
Supplier:   SWED HANDLING
Potassium nitrate
Catalog # SWEDKA580412-5
Supplier:  SWED HANDLING
CAS Number:  7757-79-1
Potassium nitrate

Plastic bottle for solids, 5 kg

CAS Number:  7757-79-1
Price
kr 1 775,00
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Melting Pt:334 °C
  • Density:2.109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.