Potassium iodide ≥99.0% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Merck Index:13,07727
Boiling Pt:1.323 °C (1.013 hPa)
Melting Pt:681 °C
Density:3.12 g/cm³ (20 °C)
EINECS:231-659-4
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99.0 %
- pH (5 %, 25 °C):6.0 - 9.2
- Loss on drying:Max. 0.2 %
- Water insoluble matter:Max. 0.005 %
- Barium (Ba):Max. 0.002 %
- Calcium (Ca):Max. 0.002 %
- Iron (Fe):Max. 3 ppm
- Magnesium (Mg):Max. 0.001 %
- Sodium (Na):Max. 0.005 %
- Heavy metals (as Pb):Max. 5 ppm
- Bromide, chloride (as Cl):Max. 0.01 %
- Iodate (IO₃):Max. 3 ppm
- Phosphate (PO₄):Max. 0.001 %
- Sulfate (SO₄):Max. 0.005 %