kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Rubidium chloride
Rubidium chloride
Rubidium chloride
PROONIST-984
:
LGC Standards PROMOCHEM
:
Rubidium chloride
PROONIST-984
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
250 mg
Seq:
2590
:
:
120.92 g/mol
:
1390 °C (1013 hPa)
:
715 °C
:
2.8 g/cm³ (20 °C)
:
Ambient
:
232-240-9
:
7791-11-9
:
RbCl
:
MFCD00011187