
Cesium chloride ≥98% Grade II, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Merck Index:13,02021
Boiling Pt:1303 °C (1013 hPa)
Melting Pt:645 °C (lit.)
Density:3,988 g/cm³ (20 °C)
EINECS:231-600-2
:GHS08
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):White
- Appearance (Form):Powder
- Solubility (Colour):Colorless
- Solubility (Turbidity)
1 g/ml, H₂O:Clear - A280 UV Absorption
50% in H₂O:≤0.2 - Sulfate (SO₄):≤0.05 %
- Titration with AgNO₃:≥ 98 %
- Recommended Retest Period:4 years
Specification Test Results
Frequently Bought Together