Propylene carbonate, Supelco®
Catalog # 82227-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:102.09 g/mol
Boiling Pt:241…242 °C (1013 hPa)
Melting Pt:–48.8 °C
Density:1.2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:132 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-572-1
:GHS07
CAS Number:108-32-7
Formula:C₄H₆O₃
MDL Number:MFCD00005385